Accuform KDD131 Double-Triple Pole Circuit Breaker Lockout
Representative image only.
Accuform Mfr # KDD131 – Item # EW-40504-10
Lock out electrical power at the source – right at the circuit breaker, no need to lock the entire panel
  • Manufactured of rigid plastic in visible bright red
  • No modifications required, attach directly to the breaker toggle and panel to prevent the breaker from being switched on once padlock is in place
  • Fits over most major brands of breaker toggles, in the off position
  • Manufactured of rigid plastic in visible bright red
  • Accommodates padlocks with shackle diameter up to 9/32" (7.1 mm) (sold separately)
$33.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDouble-Triple Pole Circuit Breaker Lockout
Key Features
  • Manufactured of rigid plastic in visible bright red
  • No modifications required, attach directly to the breaker toggle and panel to prevent the breaker from being switched on once padlock is in place
  • Fits over most major brands of breaker toggles, in the off position
  • Manufactured of rigid plastic in visible bright red
  • Accommodates padlocks with shackle diameter up to 9/32" (7.1 mm) (sold separately)
  • Double/triple-pole style fits over two or three toggles connected by a cross bar
More About this Item
No need to lock out the entire panel; these compact, universally designed lockouts fit over most major brands of breaker toggles in the "off" position. Accommodates padlocks (not included) with shackle diameter up to 9/32" (7.1 mm).
Double/Triple-Pole Style fits over two to three toggles connected by a cross bar.
Call Us At1-847-549-7600
Live chat