Accuform LZH616PS2 GHS Label,Skull/Crossbones Pictogram,2" x 2",adhesive paper,250/roll
Representative image only.
Accuform Mfr # LZH616PS2 – Item # EW-50712-92
$20.50USD / Roll of 250
SPECS & DESCRIPTION
Specifications & Description
  • HeaderSKULL/CROSSBONES
  • LanguagesEnglish
  • MaterialAdhesive Paper
  • Width (in)2
  • Height (in)2
  • Quantity250/roll
  • DescriptionGHS Label,Skull/Crossbones Pictogram,2" x 2",adhesive paper,250/roll
More About this Item
GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Skull and Crossbones pictogram indicates the following potential hazard: acute toxicity (fatal or toxic).