Accuform MRDM102VA ANSI Sign, Danger-Do Not Enter Authorized Only, 7" x 10", Aluminum
Representative image only.
Accuform Mfr # MRDM102VA – Item # EW-50712-08
Use this sign to prevent unauthorized entry
SPECS & DESCRIPTION
Specifications & Description
  • HeaderDANGER
  • LanguagesEnglish
  • MaterialAluminum
  • Width (in)10
  • Height (in)7
  • DescriptionANSI Sign, Danger-Do Not Enter Authorized Only, 7\" x 10\", Aluminum
More About this Item
ANSI-style signs include a visual alert symbol (triangle with an exclamation point), a large symbol, and a hazard message. The large symbol gives a quick and easily understand indication of the potential hazard. Recognized by OSHA as an acceptable safety notification device.
ANSI Danger Signs (Red) indicate a hazardous situation which, if not avoided, will result in death or serious injury.