Accumet CP620130 accuCap pH Probe, Refillable/DJ/Glass/160x12mm; BNC
Representative image only.
Accumet Mfr # CP620130 – Item # EW-55500-50
Most accurate pH electrode, as tested by the PTB (the German Federal Physical Technical Institute in Braunschweig)*
  • Precise, reliable, and fast readings
  • About 200 times larger junction than standard electrodes makes it impossible to clog
PRODUCT OPTIONS
$502.35USD / Each
$591.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsRefillable
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Reference JunctionSingle pore, capillary
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • ConnectorBNC
  • DescriptionaccuCap pH Probe, Refillable/DJ/Glass/160x12mm; BNC
Key Features
  • Precise, reliable, and fast readings
  • About 200 times larger junction than standard electrodes makes it impossible to clog
More About this Item
The accuCap electrodes use a single pore capillary reference junction, about 200 times larger than typical ceramic junctions. The unique junction design provides increased electrolyte flow, resulting in greater contact between sample and reference, ensuring fast, stable readings. The double junction design offers enhanced performance. A diffusion barrier separates the AgCl reservoir from the reference electrolyte, allowing the use of silver-free electrolytes and preventing AgCl loss due to temperature variations. The end result is an extremely stable reference potential, providing long electrode life.
*See "Traceability of pH measurement" by Petra Spitzer; ISBN 3-89429-877-4 or SSN 0947-7063
accuCap Refillable Electrode
Unique fluid-gel electrolyte and accuCap junction design leads to a faster flow rate and improved contact between the reference electrode and sample. It performs well in aggressive samples, liquids with low ion concentration, or partly aqueous samples.