ACL 6002 Staticide , Mat and Table Top Cleaner, 1 Gallon
Representative image only.
ACL Mfr # 6002 – Item # EW-26814-25
Safely and effectively removes surface dirt on workstations and antistatic mats
  • Safe to use on any surface and cleans without leaving any film residue
  • Enhances surface resistivity
  • Ideal for cleaning production areas used for assembling, and testing
  • Nonflammable, nonabrasive; contains no ammonia or alkali
  • Removes flux residues, oil, finger marks, grease, nicotine, and inorganic contaminants
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionStaticide , Mat and Table Top Cleaner, 1 Gallon
  • Warranty1 Year
1 Year Warranty
Key Features
  • Safe to use on any surface and cleans without leaving any film residue
  • Enhances surface resistivity
  • Ideal for cleaning production areas used for assembling, and testing
  • Nonflammable, nonabrasive; contains no ammonia or alkali
  • Removes flux residues, oil, finger marks, grease, nicotine, and inorganic contaminants