Acros Organics 4-Aminoantipyrine, 98%; 100 g

Representative image only.
Acros Organics Mfr # AC103151000 – Item # EW-88089-79
Reagents and solutions to meet your lab and research needs
  • Quality and consistency your lab can count on

$149.00USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC11H13N3O
  • ClassSolids
  • ChemistryAnalytical Reagents;Solutions
  • Synonyms4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one; 4-Aminophenazone; Ampyrone
  • FormSolid
  • Weight (g)100
  • Formula Weight203.25
  • PackagingGlass bottle
  • DOT InformationNot regulated by DOT
  • Hazard SymbolHarmful
  • CAS Number83-07-8
  • Description4-Aminoantipyrine 98% (100g)
Harmful

Key Features

  • Quality and consistency your lab can count on

More About this Item

Quality and consistency your lab can count on.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat