Acros Organics AC107451000 N-Bromosuccinimide 99% (100g) CAS 128-08-5

Representative image only.
Acros Organics Mfr # AC107451000 – Item # EW-88096-27

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC4H4BrNO2
  • ClassSolids
  • ChemistrySolutions
  • FormSolid
  • Synonyms1-Bromo-2,5-pyrolidinedione, NBS
  • Weight (g)100
  • Formula Weight177.99
  • PackagingGlass
  • DOT InformationCorrosive Solids, n.o.s.; Class 8, Packing Group III
  • Hazard SymbolHarmful
  • CAS Number128085
  • DescriptionN-Bromosuccinimide 99% (100g) CAS 128-08-5
Harmful

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat