Acros Organics AC112835000 Dibromomethane 99% (500 mL) CAS 74-95-3 (Methylene bromide)

Representative image only.
Acros Organics Mfr # AC112835000 – Item # EW-88104-54
Consistent, high quality organics and fine chemicals at great savings

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaCH2Br2
  • Linear FormulaCH2Br2
  • ClassReagent Solvents
  • ChemistrySolutions
  • FormLiquid
  • SynonymsMethylene bromide
  • Volume (mL)500
  • Formula Weight173.83
  • DOT InformationDibromoethane; Class 6.1, Packing Group III
  • Hazard SymbolHarmful
  • CAS Number74953
  • UN NumberUN2664
  • DescriptionDibromomethane 99% (500 mL) CAS 74-95-3 (Methylene bromide)
Harmful

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat