Acros Organics AC113062500 Dichloroacetic Acid, 99+%; 250 mL

Representative image only.
Acros Organics Mfr # AC113062500 – Item # EW-88105-01
Consistent, high quality organics and fine chemicals at great savings

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC2H2Cl2O2
  • Linear FormulaCl2CHCO2H
  • ClassReagent Acids and Bases
  • ChemistryAcids;Inorganics
  • FormLiquid
  • Volume (mL)250
  • Formula Weight128.94
  • PackagingGlass
  • DOT InformationDichloroacetic acid; Class 8, Packing Group II
  • Hazard SymbolCorrosive, Environment
  • CAS Number79436
  • UN NumberUN1764
  • DescriptionDichloroacetic acid 99+% (250 mL) CAS 79-43-6
Corrosive

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat