Acros Organics AC115850050 Dansyl Chloride, 98%; 5 g

Representative image only.
Fisher Chemical Mfr # AC115850050 – Item # EW-88109-66
Reagents and solutions to meet your lab and research needs
  • Quality and consistency your lab can count on

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC12H12ClNO2S
  • ClassSolids
  • ChemistrySolutions
  • Synonyms5-Dimethylamino-1-naphthalenesulfonyl chloride
  • FormSolid
  • Weight (g)5
  • Formula Weight269.75
  • PackagingGlass bottle
  • DOT InformationWATER-REACTIVE SOLIDS, CORROSIVE, N.O.S.; Class 4.3 (8), Packing Group I
  • Hazard SymbolCorrosive, Flammable, Reactive, Toxic
  • CAS Number605-65-2
  • UN NumberUN3131
  • DescriptionDansyl chloride 98% (5g)
Irritant

Key Features

  • Quality and consistency your lab can count on

More About this Item

Quality and consistency your lab can count on.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat