Acros Organics AC123350025 Calcium chloride dihydrate, flakes for drying purposes 77-80% CaCl2 (2.5kg)

Representative image only.
Acros Organics Mfr # AC123350025 – Item # EW-88122-05

$169.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaCaCl2•2H2O
  • Linear FormulaCaCl2•2H2O
  • ClassReagent Salts
  • ChemistrySolutions
  • FormFlakes
  • SynonymsNone known.
  • Weight (g)2500
  • Formula Weight147.02
  • PackagingPlastic drum
  • DOT InformationNot regulated by DOT
  • Hazard SymbolToxic
  • CAS Number10035-04-8
  • DescriptionCalcium Chloride Dihydrate, flakes for drying purposes 77-80% CaCl2 (2.5 kg)

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat