Acros Organics AC125430100 Mercaptoacetic acid 98% (10ml)

Representative image only.
Acros Organics Mfr # AC125430100 – Item # EW-88124-54

SPECS & DESCRIPTION

Specifications & Description

  • ChemistryInorganics;Acids
  • Volume (mL)10
  • Formula Weight92.11
  • CAS Number68-11-1
  • DescriptionMercaptoacetic acid 98% (10ml)

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat