Acros Organics Methyl 4-hydroxybenzoate 99% (100g)

Representative image only.
Acros Organics Mfr # AC126961000 – Item # EW-88127-28
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs

$59.10USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC8H8O3
  • Linear FormulaHOC6H4CO2CH3
  • ClassSolids
  • ChemistrySolutions
  • FormSolid
  • SynonymsMethylparaben
  • Weight (g)100
  • Formula Weight152.15
  • PackagingPlastic bottle
  • DOT InformationNot regulated by DOT
  • CAS Number99-76-3
  • DescriptionMethyl 4-hydroxybenzoate 99% (100g)

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat