Acros Organics Propylene Carbonate, 99.5%; 2.5 L

Representative image only.
Acros Organics Mfr # AC131560025 – Item # EW-88134-57
Reagents and solutions to meet your lab and research needs
  • Quality and consistency your lab can count on

PRODUCT OPTIONS

$426.50USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity Grade99.5%
  • ChemistrySolutions
  • Weight (g)2500
  • Weight (mL)2500
  • Formula Weight102.09
  • CAS Number108-32-7
  • DescriptionPropylene carbonate 99.5% (2.5kg)
Irritant

Key Features

  • Quality and consistency your lab can count on

More About this Item

Quality and consistency your lab can count on.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat