Acros Organics AC132195000 Quinoline 96% (500 mL) CAS 91-22-5 (Benzo[b]pyridine)

Representative image only.
Acros Organics Mfr # AC132195000 – Item # EW-88135-40

$94.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC9H7N
  • ClassReagent Solvents
  • ChemistrySolutions
  • FormLiquid
  • SynonymsBenzo[b]pyridine
  • Volume (mL)500
  • Formula Weight129.16
  • PackagingGlass bottle
  • DOT InformationQuinoline; Class 6.1, Packing Group III
  • Hazard SymbolFlammable, Toxic
  • UN NumberUN2656
  • CAS Number91-22-5
  • DescriptionQuinoline 96% (500 mL) CAS 91-22-5 (Benzo[b]pyridine)

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat