Acros Organics AC132641000 Semicarbazide hydrochloride p.a. (100g)

Representative image only.
Acros Organics Mfr # AC132641000 – Item # EW-88135-87

SPECS & DESCRIPTION

Specifications & Description

  • ChemistrySolutions
  • Weight (g)100
  • Formula Weight111.53
  • CAS Number563-41-7
  • DescriptionSemicarbazide hydrochloride p.a. (100g)

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat