Acros Organics AC133400250 Salicylsalicylic acid 99% (25g) CAS 552-94-3

Representative image only.
Acros Organics Mfr # AC133400250 – Item # EW-88136-61

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC14H10O5
  • ClassReagent Acids and Bases
  • ChemistryAcids
  • FormSolid
  • Synonyms2-Hydroxybenzoic acid 2-carboxyphenyl ester
  • Weight (g)25
  • Formula Weight258.22
  • PackagingGlass bottle
  • DOT InformationNot regulated by DOT
  • Hazard SymbolHarmful
  • CAS Number552-94-3
  • DescriptionSalicylsalicylic acid 99% (25g) CAS 552-94-3

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat