Acros Organics AC157741000 Sarcosine 98% (100g) CAS 107-97-1 (N-Methylglycine)

Representative image only.
Acros Organics Mfr # AC157741000 – Item # EW-88163-35

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC3H7NO2
  • Linear FormulaCH3NHCH2CO2H
  • ClassSolids
  • ChemistryAnalytical Reagents;Solutions
  • FormCrystalline powder
  • SynonymsN-Methylglycine
  • Weight (g)100
  • Formula Weight89.09
  • PackagingPlastic bottle
  • DOT InformationNot regulated by DOT
  • CAS Number107-97-1
  • DescriptionSarcosine 98% (100g) CAS 107-97-1 (N-Methylglycine)

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat