Acros Organics AC157745000 Sarcosine 98% (500g) 8816336

Representative image only.
Acros Organics Mfr # AC157745000 – Item # EW-88163-36

SPECS & DESCRIPTION

Specifications & Description

  • ChemistryAnalytical Reagents;Solutions
  • Formula Weight89.09
  • CAS Number107-97-1
  • DescriptionSarcosine 98% (500g) 8816336

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat