Acros Organics AC165015000 Methyl myristate 99% (500 mL) CAS 124-10-7

Representative image only.
Acros Organics Mfr # AC165015000 – Item # EW-88173-97

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC15H30O2
  • Linear FormulaCH3(CH2)12CO2CH3
  • ClassSolids
  • ChemistrySolutions
  • FormLiquid
  • SynonymsMethyl tetradecanoate
  • Volume (mL)500
  • Formula Weight242.4
  • PackagingGlass bottle
  • DOT InformationNot regulated by DOT
  • CAS Number124-10-7
  • DescriptionMethyl myristate 99% (500 mL) CAS 124-10-7

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat