Acros Organics AC167295000 Myristic acid 99.5+% (500g)

Representative image only.
Acros Organics Mfr # AC167295000 – Item # EW-88177-31

SPECS & DESCRIPTION

Specifications & Description

  • ChemistryAcids
  • Formula Weight228.37
  • CAS Number544-63-8
  • DescriptionMyristic acid 99.5+% (500g)

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat