Acros Organics Silicone Oil, High Temperature; 500 g
Representative image only.
Acros Organics Mfr # AC174665000 – Item # EW-88187-36
Improve performance of your lab equipment
  • Reagents and solutions to meet your lab and research needs
SPECS & DESCRIPTION
Specifications & Description
  • ChemistrySolutions
  • Weight (g)500
  • CAS Number63148-58-3
  • DescriptionSilicone oil, high temperature (500g)
Key Features
  • Reagents and solutions to meet your lab and research needs
More About this Item
Silicone oils have thermal stability, electrical insulation, and lubrication properties. Their temperature stability and good heat-transfer characteristics make them ideal for use in laboratories for heating baths placed on top of hotplate stirrers, as well as in freeze-dryers as refrigerants.