Acros Organics AC196311000 Sodium atomic absorption standard solution 1 mg/ml Na in 0.5N HNO3 (100ml)
Representative image only.
Acros Organics Mfr # AC196311000 – Item # EW-88209-40
1000 ppm : Sodium Carbonate in 2% Nitric Acid Standardized and Verified against NIST Standard Reference Material No. 3152
SPECS & DESCRIPTION
Specifications & Description
  • ChemistrySolutions
  • Volume (mL)100
  • DescriptionSodium atomic absorption standard solution 1 mg/ml Na in 0.5N HNO3 (100ml)
Irritant