Acros Organics AC197680250 Silver nitrate p.a. (25g)

Representative image only.
Ricca Chemical Company Mfr # AC197680250 – Item # EW-88211-43

SPECS & DESCRIPTION

Specifications & Description

  • ChemistrySolutions
  • Formula Weight169.87
  • CAS Number7761-88-8
  • DescriptionSilver nitrate p.a. (25g)

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat