Acros Organics AC201761000 Sodium iodate p.a. (100g) CAS 7681-55-2

Representative image only.
Acros Organics Mfr # AC201761000 – Item # EW-88215-55

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradePro Analysi
  • Empirical FormulaINaO3
  • Linear FormulaNaIO3
  • ClassReagent Salts
  • ChemistrySolutions
  • FormCrystalline powder
  • Weight (g)100
  • Formula Weight197.89
  • PackagingPlastic bottle
  • DOT InformationOXIDIZING SOLID, N.O.S.; Class 5.1, Packing Group II
  • Hazard SymbolOxidizing
  • CAS Number7681-55-2
  • UN NumberUN1479
  • DescriptionSodium iodate p.a. (100g) CAS 7681-55-2
Oxidizing

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat