Acros Organics AC202105000 Zinc bromide, anhydrous 98+% (500g) CAS 7699-45-8

Representative image only.
Acros Organics Mfr # AC202105000 – Item # EW-88215-96
  • Insoluble in Dilute HCl - ≤0.005%
  • Iron - ≤0.001%
  • Lead - ≤0.005%
  • Alkalis and Earths - ≤0.2%
  • Sulfate - ≤0.01%

PRODUCT OPTIONS

$184.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaBr2Zn
  • Linear FormulaZnBr2
  • ClassReagent Salts
  • ChemistryAnalytical Reagents;Solutions
  • FormSolid
  • Weight (g)500
  • Formula Weight225.19
  • Hazard SymbolIrritant
  • CAS Number7699-45-8
  • DescriptionZinc bromide, anhydrous 98 % (500g) CAS 7699-45-8

Key Features

  • Insoluble in Dilute HCl - ≤0.005%
  • Iron - ≤0.001%
  • Lead - ≤0.005%
  • Alkalis and Earths - ≤0.2%
  • Sulfate - ≤0.01%

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat