Acros Organics AC203180010 Sodium iodide, anhydrous 99+% (1kg)

Representative image only.
Acros Organics Mfr # AC203180010 – Item # EW-88217-16
Consistent, high quality organics and fine chemicals at great savings
  • Reagents and solutions to meet your lab and research needs

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaINa
  • Linear FormulaNaI
  • ClassReagent Salts
  • ChemistrySolutions
  • FormCrystalline
  • Weight (g)1000
  • Formula Weight149.89
  • PackagingGlass bottle
  • DOT InformationEnvironmentally Hazardous Substances, Solid, N.O.S.; Class 9, Packing Group III
  • Hazard SymbolEnvironmental, Harmful
  • CAS Number7681-82-5
  • UN NumberUN3077
  • DescriptionSodium iodide, anhydrous 99+% (1kg)

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat