Acros Organics AC206520010 Sodium phosphate, tribasic dodecahydrate p.a. (1kg)

Representative image only.
Acros Organics Mfr # AC206520010 – Item # EW-88221-10

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaNa3O4P•12H2O
  • Linear FormulaNa3PO4•12H2O
  • ClassReagent Salts
  • ChemistrySolutions
  • FormCrystalline
  • SynonymsTrisodium phosphate dodecahydrate; TSP
  • Weight (g)1000
  • Formula Weight380.13
  • PackagingPlastic bottle
  • DOT InformationNot regulated by DOT
  • Hazard SymbolHarmful
  • CAS Number10101-89-0
  • DescriptionSodium phosphate, tribasic dodecahydrate p.a. (1kg)
Irritant

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat