Acros Organics AC207621000 Sodium tungstate dihydrate p.a. (100g) CAS 10213-10-2

Representative image only.
Fisher Chemical Mfr # AC207621000 – Item # EW-88222-25
  • Assay - 99.0-101.0%
  • Insoluble Matter - ≤0.01%
  • Chloride - ≤0.005%
  • Molybdenum - ≤0.001%
  • Nitrogen Compounds (as N) - ≤0.001%
  • Sulfate - ≤0.01%
  • Heavy Metals and Iron (as Pb) - ≤0.001%
  • Titratable Free Base - ≤0.02mEq/g

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradePro Analysi
  • Empirical FormulaNa2O4W•2H2O
  • Linear FormulaNa2WO4•2H2O
  • ClassSolids
  • ChemistrySolutions
  • FormFlakes
  • Weight (g)100
  • Formula Weight329.85
  • PackagingGlass bottle
  • DOT InformationNot regulated by DOT
  • Hazard SymbolHarmful
  • CAS Number10213-10-2
  • DescriptionSodium tungstate dihydrate p.a. (100g) CAS 10213-10-2
Harmful

Key Features

  • Assay - 99.0-101.0%
  • Insoluble Matter - ≤0.01%
  • Chloride - ≤0.005%
  • Molybdenum - ≤0.001%
  • Nitrogen Compounds (as N) - ≤0.001%
  • Sulfate - ≤0.01%
  • Heavy Metals and Iron (as Pb) - ≤0.001%
  • Titratable Free Base - ≤0.02mEq/g

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat