Acros Organics Sodium Peroxide, p.a.; 25 g

Representative image only.
Acros Organics Mfr # AC207700250 – Item # EW-88222-40
Reagents and solutions to meet your lab and research needs
  • Quality and consistency your lab can count on

$90.10USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradePro Analysi
  • Empirical FormulaNa2O2
  • Linear FormulaNa2O2
  • ClassReagent Salts
  • ChemistrySolutions
  • FormPowder
  • Weight (g)25
  • Formula Weight77.97
  • DOT InformationSodium Peroxide; Class 5.1, Packing Group I
  • Hazard SymbolCorrosive, Oxidizing
  • UN NumberUN1504
  • CAS Number1313-60-6
  • DescriptionSodium peroxide p.a. (25g) CAS 1313-60-6
CorrosiveOxidizing

Key Features

  • Quality and consistency your lab can count on

More About this Item

Quality and consistency your lab can count on.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat