Acros Organics Drierite™ Desiccant Without Indicator, 8 Mesh (500g)

Representative image only.
Acros Organics Mfr # AC219055000 – Item # EW-88234-13
Keeps samples dry and moisture free
  • Reagents and solutions to meet your lab and research needs

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaCaO4S
  • ClassSolids
  • FormSolid
  • Weight (g)500
  • Formula Weight136.14
  • PackagingGlass bottle
  • DOT InformationNot regulated by DOT
  • CAS Number7778-18-9
  • DescriptionDrierite™ Desiccant Without Indicator, 8 Mesh (500g)

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

These desiccants are designed for use in laboratory settings and are commonly used to keep substances and samples dry, and to prevent moisture from forming on and within containers.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat