Acros Organics AC219240010 Sodium dichromate dihydrate 99% (1kg) CAS 7789-12-0

Representative image only.
Acros Organics Mfr # AC219240010 – Item # EW-88234-50

$155.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaCr2Na2O7•2H2O
  • Linear FormulaNa2Cr2O7•2H2O
  • ClassReagent Salts
  • ChemistrySolutions
  • FormCrystalline
  • SynonymsSodium bichromate
  • Weight (g)1000
  • Formula Weight298
  • PackagingPlastic bottle
  • DOT InformationOXIDIZING SOLID, TOXIC, N.O.S.; Class 5.1 (6.1), Packing Group II
  • Hazard SymbolEnvironment, Highly Toxic
  • UN NumberUN3087
  • CAS Number7789-12-0
  • DescriptionSodium dichromate dihydrate 99% (1kg) CAS 7789-12-0

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat