Acros Organics AC270540010 Sodium oxalate p.a. (1kg)

Representative image only.
Acros Organics Mfr # AC270540010 – Item # EW-88260-85
Quality and consistency your lab can count on

SPECS & DESCRIPTION

Specifications & Description

  • ChemistrySolutions
  • Weight (g)1000
  • Formula Weight134
  • CAS Number62-76-0
  • DescriptionSodium oxalate p.a. (1kg)
Harmful

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat