Acros Organics AC424425000 Sodium sulfide nonahydrate, reagent ACS, crystals (500g)

Representative image only.
Acros Organics Mfr # AC424425000 – Item # EW-88363-06
  • Ammonium - ≤0.01%
  • Sulfite and Thiosulfate (as SO2) - To pass test (about 2%)
  • Iron - To pass test

$169.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • ChemistryAnalytical Reagents;Solutions
  • Weight (g)500
  • Formula Weight240.19
  • CAS Number1313-84-4
  • DescriptionSodium sulfide nonahydrate, reagent ACS, crystals (500g)

Key Features

  • Ammonium - ≤0.01%
  • Sulfite and Thiosulfate (as SO2) - To pass test (about 2%)
  • Iron - To pass test

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat