Acros Organics Ammonium Persulfate, 98+%, Reagent ACS; 500 g

Representative image only.
Acros Organics Mfr # AC401165000 – Item # EW-88338-96
Reagents and solutions to meet your lab and research needs
  • Quality and consistency your lab can count on

PRODUCT OPTIONS

$196.75USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • ChemistrySolutions
  • Weight (g)500
  • Formula Weight228.2
  • CAS Number7727-54-0
  • DescriptionAmmonium persulfate, reagent ACS 98+% (500g)
HarmfulOxidizing

Key Features

  • Quality and consistency your lab can count on

More About this Item

Quality and consistency your lab can count on.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat