Acros Organics Water, HPLC Gradient Grade, 4 L

Representative image only.
Acros Organics Mfr # AC268300040 – Item # EW-88258-89
Quality and consistency your lab can count on
  • Water of various purity grades suitable for an array of applications
  • Ultra-pure water suitable for HPLC processes and specially prepared to have low UV-absorptivity

PRODUCT OPTIONS

$191.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeHPLC Gradient
  • Empirical FormulaH2O
  • Synonymsdihydrogen oxide, dihydrogen monoxide
  • FormLiquid
  • Volume (mL)4000
  • Viscosity (cSt)0.89
  • Formula Weight18.02
  • DOT InformationNot regulated by DOT
  • CAS Number7732-18-5
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • DescriptionWater, HPLC Gradient Grade, 4 L

Key Features

  • Water of various purity grades suitable for an array of applications
  • Ultra-pure water suitable for HPLC processes and specially prepared to have low UV-absorptivity

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat