Acros Organics Zinc Bromide, Anhydrous 98+%; 100 g

Representative image only.
Acros Organics Mfr # AC202101000 – Item # EW-88215-95
Reagents and solutions to meet your lab and research needs
  • Quality and consistency your lab can count on

PRODUCT OPTIONS

$149.75USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaBr2Zn
  • Linear FormulaZnBr2
  • ClassReagent Salts
  • ChemistryAnalytical Reagents;Solutions
  • FormPowder
  • Weight (g)100
  • Formula Weight225.19
  • PackagingGlass bottle
  • DOT InformationCORROSIVE SOLIDS, ACIDIC, INORGANIC, N.O.S.; Class 8, Packing Group II
  • Hazard SymbolCorrosive, Toxic
  • UN NumberUN3260
  • CAS Number7699-45-8
Irritant

Key Features

  • Quality and consistency your lab can count on

More About this Item

Quality and consistency your lab can count on.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat