Eurotherm T713-0000 RTD Transmitterintrinsically Safe, 4-20 Ma Out
Representative image only.
Eurotherm Mfr # T713-0000 – Item # EW-68478-19
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Control Input RTDYes
  • Loop Power SupplyYes
  • DescriptionRTD Transmitterintrinsically Safe, 4-20 Ma Out
  • Warranty3 Years
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
These two-wire transmitters are compact, rugged units that receive signals and direct sensor inputs, and operate from power derived from the output signal loop. This series provides a complete line of direct interface and other process parameters. These transmitters also accept a wide range of other electrical DC and AC voltage and current inputs.