Adam Equipment 1140013826 Roller Deck, 11.8" x 15.7"
Representative image only.
Adam Equipment Mfr # 1140013826 – Item # UX-11119-76
Provides easy handling of boxes and crates
  • For use with AGB and AGF industrial scales
  • 2 year warranty
PRODUCT OPTIONS
$249.37USD / Each
$285.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypeRoller deck
  • Height (in)2 1/2
  • Width (in)11 51/64
  • Length (in)15 45/64
  • Warranty2 year
Two year warranty
Key Features
  • For use with AGB and AGF industrial scales
  • 2 year warranty
More About this Item
These heavy duty stainless steel roller decks are designed to fit in place of the existing pan and allows boxes or crates to pass easily onto it for weighing tasks. Built-in handles allow for easy placement on scales.