Additel ADT100-761-4N Hose Kit for ADT761 Automated Pressure Calibrator
Representative image only.
Additel Mfr # ADT100-761A-N4 – Item # EW-16106-71
Convenient for fast and tool-free calibrations
PRODUCT OPTIONS
$489.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHose Kit for ADT761 Automated Pressure Calibrator
More About this Item
Each component is precision machined so that leak-free connections can be made by hand—no tools or tape necessary!
5-ft of 4-mm ID flexible hose, and 1/64" tube to 1/4"NPT(F) hand-tight quick connector.