Additel ADT100-HTK-NPTM4-NPTF4Q Pressure Hose; 1/4"NPTM, 1/4"NPTF Quick-Connect
Representative image only.
Additel Mfr # ADT100-HTK-15K-NPTM4-BSPF4Q – Item # EW-16108-23
Convenient for fast and tool-free calibrations
  • 1 year warranty
PRODUCT OPTIONS
$306.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPressure Hose; 1/4\"NPTM, 1/4\"NPTF Quick-Connect
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Each component is precision machined so that leak-free connections can be made by hand—no tools or tape necessary!
Hose test kit includes 5 ft (1.5 m) flexible hose (15,000 psi) and 1/4" NPT(M) to 1/4" NPT(F) hand-tight quick connector.