Advantec 311580 Glass Microanalysis Filter Holder, 47 mm, SS Support; 100 mL
Representative image only.
Advantec Mfr # 311580 – Item # EW-06644-89
Ideal for small to medium volumes of liquid
  • Standard sizes for microbiology and particulate analysis
  • Select glass support for low pH solutions or for slower flow rates; use stainless for proteinaceous or viscous solutions; use PTFE support for acidic solutions at higher flow rates
PRODUCT OPTIONS
$504.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter Diameter (mm)47
  • Body MaterialPTFE
  • Funnel Size (mL)100
  • Filtration Area (cm²)9.6
  • SupportStainless Steel
  • Stopper SizeNo. 8
  • DescriptionGlass Microanalysis Filter Holder, 47 mm, SS Support; 100 mL
Key Features
  • Standard sizes for microbiology and particulate analysis
  • Select glass support for low pH solutions or for slower flow rates; use stainless for proteinaceous or viscous solutions; use PTFE support for acidic solutions at higher flow rates
More About this Item
Microanalysis filter holders feature borosilicate glass funnel and base, silicone rubber stopper, and anodized aluminum clamp. The 90-mm holders are ideal for large-volume viscous samples or those with a heavy particle or bioburden that would clog a 47-mm filter.