This item is now discontinued. We recommend the following as suggested replacements: EW-60060-89
Advantec 370S2000N Air monitoring cassette, 2-piece, 37-mm diameter; pack of 50
Representative image only.
Advantec Mfr # 370S2000N – Item # EW-02935-30
Assemble you own monitor for specific applications
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Filter Diameter (mm)37
  • Body Material Styrene Acrylonitrile
  • DescriptionAir monitoring cassette, 2-piece, 37-mm diameter; pack of 50
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Empty cassettes and cellulose pads are available as an option for you to assemble a filter/monitoring unit with an MCE membrane filter for asbestos monitoring or other 37-mm filter.