Advantec 501015 Silicone O-Ring for Polysulfone Filter System Receiver Flask
Representative image only.
Advantec Mfr # 501015 – Item # EW-06645-94
Enhance the life of your Advantec Polysulfone filter holders
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • O-RingSilicone
  • DescriptionSilicone O-Ring for Polysulfone Filter System Receiver Flask
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
These parts and accesories are suitable for use with your Advantec polysulfone filter holders.
The funnel cover can be vented aseptically using disposable syringe filter.
The recessed filter support provides sufficient clearance so that the membrane will not twist or tear when the funnel is secured.
Aseptic Polysulfone Filter Holder sold separately as part numbers 06644-70, -72, -74, and -76.