Advantec A045G047J Sterile MCE Hydrophobic Edge Filters, 0.45um, 47mm Dia; 100/Pk
Representative image only.
Advantec Mfr # A045G047J – Item # EW-06644-28
Presterilized membranes in convenient single-, multi-, or continuous-feed packs
PRODUCT OPTIONS
$182.00USD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • MaterialMixed Cellulose Ester
  • Pore Size (µm)0.45
  • Diameter (mm)47
  • SurfacePlain
  • Absorbent PadNot included
  • DescriptionSterile MCE Hydrophobic Edge Filters, 0.45um, 47mm Dia; 100/Pk
More About this Item
Mixed cellulose ester membranes are suitable for microbiological analysis of water, wastewater, pharmaceuticals, or beverages. Offered with or without grid lines; grid lines facilitate counting colonies on the membrane surface. Grid squares measure 3.1 mm and represent 1/100 of the area of a 47-mm membrane. Black membranes maximize contrast between colonies and the filter; green membrane enables viewing of black, white, and colorless colonies on a single filter.
Presterilized by EtO, these 47-mm diameter filters are available in envelopes of 10, individually packaged, or now individually wrapped continuous feed. Order membranes with absorbent pad for use with broth media.