Advantec GB100R142MM Grade GB100R Glass Fiber Filters; 142mm dia; 100/box
Representative image only.
Advantec Mfr # GB100R142MM – Item # EW-06657-77
For analytical or gravimetric studies
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass Microfiber
  • Diameter (mm)142
  • Diameter (cm)14.2
  • DescriptionGrade GB100R Glass Fiber Filters; 142mm dia; 100/box
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Low trace metal content; sampling high and low volume aerosols for airborne dust and metal contaminants.