Advantec GC50293MM Grade GC50 Glass Fiber Filters; 293mm dia; 50/box

Representative image only.
Advantec Mfr # GC50293MM – Item # EW-06657-28
For analytical or gravimetric studies

PRODUCT OPTIONS

$481.00USD / Pkg of 50

SPECS & DESCRIPTION

Specifications & Description

  • MaterialGlass Microfiber
  • Pore Size (micron)0.5
  • Diameter (mm)293
  • Diameter (cm)29.3
  • DescriptionGrade GC50 Glass Fiber Filters; 293mm dia; 50/box

More About this Item

Membrane prefilter (<0.45 µm), for industrial and wastewater analysis and scintillation counting.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat