Advantech L3-M3 Precision Sieves, Nickel Mesh, 0.003 mm Sieve Opening, 3" dia
Representative image only.
Advantech Mfr # L3-M3 – Item # EW-59948-83
  • 1 year warranty
PRODUCT OPTIONS
$1,740.00USD / Each
$157.00 - $230.00USD / Each
for an additional
$157.00USD / Each
for an additional
$230.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialAcrylic frame, electroformed nickel wire
  • Sieve Opening (mm)0.003
  • Outside Diameter (in)3
  • Height (InchShort)1.25
  • DescriptionPrecision Sieves, Nickel Mesh, 0.003 mm sieve opening
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Precision sieves feature electroformed nickel mesh. Sieves meet ASTM E161 and ISO specifications. Sieves measure 3" ID x 1-1/4" H.