Advantech L3-S200 Standard Sieve, 3" dia, Stainless Steel Mesh, 0.075 mm Sieve Opening, No. 200
Representative image only.
Advantech Mfr # L3-S200 – Item # EW-59948-45
Steel mesh meets ASTM and ISO specifications
  • 1 year warranty
PRODUCT OPTIONS
$173.50USD / Each
$157.00 - $230.00USD / Each
for an additional
$157.00USD / Each
for an additional
$230.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialAcrylic frame, stainless steel wire
  • U.S. Standard Screen Size (No.)200
  • Tyler Standard Screen Size (mesh)200
  • Sieve Opening (mm)0.074
  • Inside Diameter (InchShort)3
  • Height (InchShort)0.625
  • DescriptionStandard Sieve, Stainless Steel Mesh, 0.075 mm sieve opening, No. 200
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Standard sieves with stainless steel mesh meet ASTM and ISO specifications and are supplied with a certificate of compliance. Sieves measure 3" ID and 5/8" H.