Advantech L3-S230 Standard Sieve, Stainless Steel Mesh, 0.063 mm sieve opening, No. 230
Representative image only.
Advantech Mfr # L3-S230 – Item # EW-59948-46
$145.60USD / Each
$65.00 - $133.00USD / Each
for an additional
$65.00USD / Each
for an additional
$133.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialAcrylic frame, stainless steel wire
  • U.S. Standard Screen Size (No.)230
  • Tyler Standard Screen Size (mesh)250
  • Sieve Opening (mm)0.063
  • Height (in)0.625
  • DescriptionStandard Sieve, Stainless Steel Mesh, 0.063 mm sieve opening, No. 230
More About this Item
Standard sieves with stainless steel mesh meet ASTM and ISO specifications and are supplied with a certificate of compliance. Sieves measure 3"ID and 5/8" H.